Assay Details
Target Gene Details
Entrez Gene ID: | 407823 |
Gene Name: | bromodomain adjacent to zinc finger domain, 2B |
Gene Aliases: |
5830435C13Rik, BC053917, D2Ertd794e |
Location: |
Chr.2:59899363-60209846 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baz2b | NM_001001182.3 | NP_001001182.2 | ||
| AK029231.1 | ||||
| AK047236.1 | 2 | 516 | ||
| AK173180.1 | BAD32458.1 | |||
| BC150814.1 | ||||
| CN534605.1 |
More Information
Set Membership: |
|

Genomic Map