Assay Details
Target Gene Details
Entrez Gene ID: | 12531 |
Gene Name: | cell division cycle 25B |
Gene Aliases: |
AI604853 |
Location: |
Chr.2:131186436-131198511 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc25b | NM_001111075.4 | NP_001104545.1 | ||
| NM_023117.4 | NP_075606.1 | |||
| AK004836.1 | BAB23603.1 | |||
| AK043730.1 | BAC31632.1 | |||
| AK153903.1 | BAE32245.1 | |||
| AK157397.1 | BAE34078.1 | |||
| BC002287.1 | AAH02287.1 | |||
| BC057568.1 | AAH57568.1 | |||
| S93521.1 | AAB22026.1 |
More Information
Set Membership: |
|

Genomic Map