Assay Details
Target Gene Details
Entrez Gene ID: | 21939 |
Gene Name: | CD40 antigen |
Gene Aliases: |
AI326936, Bp50, GP39, HIGM1, IGM, IMD3, T-BAM, TRAP, Tnfrsf5, p50 |
Location: |
Chr.2:165055604-165071654 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd40 | NM_011611.2 | NP_035741.2 | ||
| NM_170702.2 | NP_733803.2 | |||
| NM_170703.2 | NP_733804.1 | |||
| NM_170704.2 | NP_733805.1 | |||
| NR_027852.1 | ||||
| AJ401387.1 | CAC29427.1 | |||
| AJ401388.1 | CAC29428.1 | |||
| AJ401389.1 | CAC29429.1 | |||
| AJ401390.1 | CAC29430.1 | |||
| AK150959.1 | BAE29991.1 | |||
| AK151458.1 | ||||
| AK152716.1 | BAE31440.1 | |||
| AK152756.1 | BAE31471.1 | |||
| AK152942.1 | BAE31613.1 | |||
| AK156644.1 | BAE33789.1 | |||
| AK161978.1 | BAE36663.1 | |||
| AK162305.1 | BAE36843.1 | |||
| BC029254.1 | AAH29254.1 | |||
| BY259264.1 | ||||
| BY743050.1 | ||||
| CT010343.1 | CAJ18551.1 | |||
| M83312.1 | AAB08705.1 |
More Information
Set Membership: |
|

Genomic Map