Assay Details
Target Gene Details
Entrez Gene ID: | 12613 |
Gene Name: | carboxyl ester lipase |
Gene Aliases: |
1810036E18Rik, BAL |
Location: |
Chr.2:28555819-28563362 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cel | NM_009885.1 | NP_034015.1 | ||
| AK131818.1 | BAE20818.1 | |||
| AK131824.1 | BAE20820.1 | |||
| BC006872.1 | AAH06872.1 | |||
| U33169.1 | AAA92088.1 | |||
| U37386.1 | AAC52279.1 |
More Information
Set Membership: |
|

Genomic Map