Assay Details
Target Gene Details
Entrez Gene ID: | 228094 |
Gene Name: | ceramide kinase-like |
Gene Aliases: |
Gm1958, Rp26 |
Location: |
Chr.2:79331105-79430520 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cerkl | NM_001048176.1 | NP_001041641.1 | ||
| CJ130836.1 | ||||
| HQ426680.1 | ||||
| HQ426681.1 | ||||
| HQ426682.1 | ||||
| HQ426683.1 | ||||
| HQ426684.1 | ||||
| HQ426685.1 | ||||
| HQ426686.1 | ||||
| HQ426687.1 | ||||
| HQ426688.1 | ||||
| HQ426689.1 | ||||
| HQ426690.1 | ||||
| HQ426702.1 | ||||
| HQ426703.1 | ||||
| HQ426704.1 | ||||
| HQ426705.1 | ||||
| HQ426706.1 | ||||
| HQ426707.1 | ||||
| HQ426708.1 | ||||
| HQ426709.1 | ||||
| HQ426710.1 | ||||
| HQ426711.1 |
More Information
Set Membership: |
|

Genomic Map