Assay Details
Target Gene Details
Entrez Gene ID: | 11435 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle) |
Gene Aliases: |
AI385656, AI608266, Achr-1, Acra |
Location: |
Chr.2:73563281-73580338 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna1 | NM_007389.5 | NP_031415.2 | ||
| AK029177.1 | BAC26337.1 | |||
| AK132589.1 | BAE21249.1 | |||
| BC125439.1 | ||||
| BC137678.1 | ||||
| X03986.1 | CAA27624.1 |
More Information
Set Membership: |
|

Genomic Map