Assay Details
Target Gene Details
Entrez Gene ID: | 11435 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 1 (muscle) |
Gene Aliases: |
AI385656, AI608266, Achr-1, Acra |
Location: |
Chr.2:73563281-73580338 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna1 | NM_007389.5 | 2 | 169 | NP_031415.2 |
| AK029177.1 | 2 | 138 | BAC26337.1 | |
| AK132589.1 | 2 | 109 | BAE21249.1 | |
| BC125439.1 | 2 | 169 | ||
| BC137678.1 | 2 | 169 | ||
| EF520684.1 | 1 | 44 | ||
| X03986.1 | 2 | 126 | CAA27624.1 |
More Information
Set Membership: |
|

Genomic Map