Assay Details
Target Gene Details
Entrez Gene ID: | 26371 |
Gene Name: | cytosolic iron-sulfur protein assembly 1 |
Gene Aliases: |
AW210570, Wdr39 |
Location: |
Chr.2:127240938-127247816 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ciao1 | NM_025296.4 | NP_079572.2 | ||
| AK002314.1 | BAB22008.1 | |||
| AK004129.1 | ||||
| AK028376.1 | BAC25915.1 | |||
| BC004089.1 | AAH04089.1 | |||
| BI734652.1 |

Genomic Map