Assay Details
Target Gene Details
Entrez Gene ID: | 66877 |
Gene Name: | Crn, crooked neck-like 1 (Drosophila) |
Gene Aliases: |
1200013P10Rik, 5730590A01Rik, C80326, crn |
Location: |
Chr.2:145917482-145934700 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crnkl1 | NM_025820.3 | NP_080096.1 | ||
| AK004749.1 | BAB23530.1 | |||
| AK012962.1 | BAB28572.1 | |||
| AK020002.1 | ||||
| AK088882.1 | BAC40630.1 | |||
| BC029187.1 | AAH29187.1 |
More Information
Set Membership: |
|

Genomic Map