Assay Details
Target Gene Details
Entrez Gene ID: | 77771 |
Gene Name: | cysteine-serine-rich nuclear protein 3 |
Gene Aliases: |
A330102K23Rik, CSRNP-3, Csnrp3, Mbu1, taip-2 |
Location: |
Chr.2:65845767-66031546 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csrnp3 | NM_001290666.1 | NP_001277595.1 | ||
NM_153409.5 | 2 | 157 | NP_700458.3 | |
NM_178634.2 | NP_848749.2 | |||
AK020730.1 | 2 | 80 | ||
AK039150.1 | 2 | 83 | BAC30257.1 | |
AK080761.1 | 2 | 80 | ||
AK082649.1 | BAC38559.1 | |||
BC055701.1 | AAH55701.1 | |||
BY297754.1 | 2 | 157 | ||
EF210820.1 | 2 | 129 |
