Assay Details
Target Gene Details
Entrez Gene ID: | 11432 |
Gene Name: | acid phosphatase 2, lysosomal |
Gene Aliases: |
Acp-2, LAP |
Location: |
Chr.2:91202812-91214098 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acp2 | NM_007387.2 | 11 | 2905 | NP_031413.1 |
| AK089401.1 | 11 | 2896 | ||
| AK128963.1 | 11 | 2898 | ||
| AK133756.1 | 11 | 2523 | BAE21825.1 | |
| AK170658.1 | 11 | 2906 |
Target Gene Details
Entrez Gene ID: | 107986 |
Gene Name: | damage specific DNA binding protein 2 |
Gene Aliases: |
2610043A19Rik |
Location: |
Chr.2:91211579-91237132 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddb2 | NM_028119.5 | NP_082395.2 | ||
| AK011756.1 | ||||
| AK158770.1 | BAE34654.1 | |||
| AY027937.1 | AAK16810.1 | |||
| BC099410.1 | AAH99410.1 | |||
| BU515611.1 |
More Information
Set Membership: |
|

Genomic Map