Assay Details
Target Gene Details
Entrez Gene ID: | 77006 |
Gene Name: | DDRGK domain containing 1 |
Gene Aliases: |
1110001I20Rik, 2600009E05Rik, AI326138, Ufbp1 |
Location: |
Chr.2:130654083-130665600 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddrgk1 | NM_029832.2 | NP_084108.1 | ||
| BC051541.1 | AAH51541.1 | |||
| BC115616.2 | ||||
| BC115617.1 |

Genomic Map