Assay Details
Target Gene Details
Entrez Gene ID: | 228889 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 27 |
Gene Aliases: |
C86129 |
Location: |
Chr.2:167015255-167034950 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx27 | NM_153065.3 | 12 | 1360 | NP_694705.2 |
AK134274.1 | 12 | 1361 | BAE22077.1 | |
AK138442.1 | 12 | 1384 | BAE23664.1 | |
AK146469.1 | 12 | 1388 | ||
BC011321.1 | 11 | 1237 | AAH11321.1 |
