Assay Details
Target Gene Details
Entrez Gene ID: | 227695 |
Gene Name: | DNA segment, Chr 2, Wayne State University 81, expressed |
Gene Aliases: |
9930028P05, AL033358 |
Location: |
Chr.2:30173447-30178495 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D2Wsu81e | NM_172660.4 | NP_766248.3 | ||
| AK036945.1 | ||||
| AK044983.1 | ||||
| AK046148.1 | BAC32613.1 | |||
| AK147162.1 | BAE27727.1 | |||
| BC078441.1 | 7 | 1448 | AAH78441.1 |
More Information
Set Membership: |
|

Genomic Map