Assay Details
Target Gene Details
Entrez Gene ID: | 69601 |
Gene Name: | disabled 2 interacting protein |
Gene Aliases: |
2310011D08Rik, AI480459, Aip1, mKIAA1743 |
Location: |
Chr.2:35558458-35730994 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dab2ip | NM_001001602.2 | 11 | 3097 | NP_001001602.2 |
| NM_001114124.2 | 13 | 3221 | NP_001107596.1 | |
| NM_001114125.1 | 13 | 3073 | NP_001107597.1 | |
| NM_001290637.1 | 13 | 3128 | NP_001277566.1 | |
| NM_001290639.1 | 11 | 2756 | NP_001277568.1 | |
| NM_001290640.1 | 11 | 3061 | NP_001277569.1 | |
| NM_001290641.1 | 13 | 3354 | NP_001277570.1 | |
| AK122548.1 | BAC65830.1 | |||
| AK147464.1 | 13 | 3153 | BAE27930.1 | |
| AK147593.1 | 13 | 3355 | BAE28013.1 | |
| AK155296.1 | 13 | 3129 | BAE33173.1 | |
| AY178784.1 | 3 | 989 | AAP31233.1 | |
| AY305656.1 | 11 | 2814 | AAQ77379.1 | |
| AY305657.1 | 11 | 3097 | AAQ77380.1 | |
| AY305658.1 | 11 | 2756 | AAQ77381.1 | |
| BC094373.1 | AAH94373.1 | |||
| BC118530.1 | 11 | 2722 | ||
| DQ473307.1 | 13 | 2958 |
More Information
Set Membership: |
|

Genomic Map