Assay Details
Target Gene Details
Entrez Gene ID: | 52840 |
Gene Name: | dysbindin (dystrobrevin binding protein 1) domain containing 2 |
Gene Aliases: |
1110017A21Rik, 2900022J10Rik, AU041050, AW048677, D2Bwg0891e, Nkip, R74724 |
Location: |
Chr.2:164472235-164493323 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dbndd2 | NM_001048227.1 | NP_001041692.1 | ||
| NM_026797.2 | NP_081073.1 | |||
| BC019947.1 | AAH19947.1 | |||
| BC030298.1 | 1 | 179 | AAH30298.1 | |
| BG080686.2 | ||||
| CX241653.1 |

Genomic Map