Assay Details
Target Gene Details
Entrez Gene ID: | 80744 |
Gene Name: | CWC22 spliceosome-associated protein homolog (S. cerevisiae) |
Gene Aliases: |
AA684037, AI173004, AL022752, B230213M24, mKIAA1604 |
Location: |
Chr.2:77895653-77946356 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cwc22 | NM_001290740.1 | 1 | 106 | NP_001277669.1 |
| NM_030560.5 | 1 | 106 | NP_085037.2 | |
| NM_172667.2 | 1 | 106 | NP_766255.1 | |
| AK132001.1 | 1 | 50 | BAE20931.1 | |
| BU613482.1 | 1 | 103 |
More Information
Set Membership: |
|

Genomic Map