Assay Details
Target Gene Details
Entrez Gene ID: | 13081 |
Gene Name: | cytochrome P450, family 24, subfamily a, polypeptide 1 |
Gene Aliases: |
24-OHase, CP24, Cyp24 |
Location: |
Chr.2:170482965-170497138 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp24a1 | NM_009996.3 | NP_034126.1 | ||
| AK143984.1 | BAE25646.1 | |||
| AK159109.1 | BAE34825.1 | |||
| AK159481.1 | BAE35118.1 | |||
| AK159527.1 | BAE35156.1 | |||
| AK159852.1 | BAE35429.1 | |||
| AK159954.1 | BAE35510.1 | |||
| AK159976.1 | BAE35528.1 | |||
| AK164697.1 | BAE37882.1 | |||
| BC109139.1 | AAI09140.1 | |||
| BC109140.1 | AAI09141.1 | |||
| D49438.1 | BAA08416.1 | |||
| D89669.1 | BAA21843.1 |
More Information
Set Membership: |
|

Genomic Map