Assay Details
Target Gene Details
Entrez Gene ID: | 13429 |
Gene Name: | dynamin 1 |
Gene Aliases: |
AI838169, Dnm, Ftfl, mKIAA4093 |
Location: |
Chr.2:32308471-32353339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnm1 | NM_001301737.1 | NP_001288666.1 | ||
| NM_010065.3 | NP_034195.2 | |||
| NR_125959.1 | ||||
| AK011651.1 | BAB27759.1 | |||
| AK144142.1 | BAE25726.1 | |||
| AK220483.1 | BAD90284.1 | |||
| BC034679.1 | AAH34679.1 | |||
| BC058623.1 | AAH58623.1 | |||
| CA328097.1 | ||||
| L31395.1 | AAA37318.1 | |||
| L31396.1 | AAA37323.1 | |||
| L31397.1 | AAA37324.1 |
More Information
Set Membership: |
|

Genomic Map