Assay Details
Target Gene Details
Entrez Gene ID: | 66861 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 10 |
Gene Aliases: |
1200006L06Rik, D2Ertd706e, ERdj5, JPDI |
Location: |
Chr.2:80315466-80354055 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc10 | NM_024181.2 | NP_077143.2 | ||
| AF255459.1 | AAN73273.1 | |||
| AF314002.1 | AAQ14555.1 | |||
| AK004617.1 | BAB23413.1 | |||
| AK028184.1 | ||||
| AK128945.1 | ||||
| AK165652.1 | ||||
| AK166502.1 | ||||
| AK169020.1 | ||||
| AK190686.1 | ||||
| AK204346.1 | ||||
| AK206826.1 | ||||
| BC002207.1 | AAH02207.1 | |||
| BC033461.1 | AAH33461.1 |
More Information
Set Membership: |
|

Genomic Map