Assay Details
Target Gene Details
Entrez Gene ID: | 110074 |
Gene Name: | deoxyuridine triphosphatase |
Gene Aliases: |
5031412I06Rik, 5133400F09Rik, D2Bwg0749e, Dutp, dUTPase |
Location: |
Chr.2:125247248-125259049 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dut | NM_001159646.1 | NP_001153118.1 | ||
| NM_023595.6 | NP_076084.2 | |||
| AF091101.1 | AAF74514.1 | |||
| AK003076.1 | BAB22551.1 | |||
| AK011407.1 | BAB27599.1 | |||
| AK017216.1 | ||||
| AK077244.1 | BAC36706.1 | |||
| AK135438.1 | BAE22532.1 | |||
| AK152459.1 | BAE31237.1 | |||
| AK153285.1 | BAE31868.1 | |||
| AK185914.1 | ||||
| AK197209.1 | ||||
| BC019979.1 | AAH19979.1 | |||
| BC053693.1 | AAH53693.1 |
More Information
Set Membership: |
|

Genomic Map