Assay Details
Target Gene Details
Entrez Gene ID: | 98221 |
Gene Name: | eukaryotic translation initiation factor 3, subunit M |
Gene Aliases: |
Ga17, Pcid1, Tango7 |
Location: |
Chr.2:104999656-105017027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3m | NM_145380.2 | NP_663355.1 | ||
AK077480.1 | BAC36821.1 | |||
AK084738.1 | BAC39270.1 | |||
AK146222.1 | BAE26990.1 | |||
AK168066.1 | BAE40042.1 | |||
BC005598.1 | AAH05598.1 | |||
BC089568.1 | AAH89568.1 | |||
BC103795.1 | AAI03796.1 | |||
BC116787.1 | ||||
BC116789.1 |
