Assay Details
Target Gene Details
Entrez Gene ID: | 16418 |
Gene Name: | eukaryotic translation initiation factor 6 |
Gene Aliases: |
1110004P11, AA408895, CAB, Itgb4bp, eIF-6, imc-415, p27(BBP), p27BBP |
Location: |
Chr.2:155819837-155826925 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif6 | NM_010579.2 | NP_034709.1 | ||
AF047046.1 | AAD28078.1 | |||
AF108462.2 | AAF07396.1 | |||
AK003447.1 | BAB22796.1 | |||
AK005384.1 | ||||
AK007594.1 | BAB25125.1 | |||
AK012218.1 | BAB28105.1 | |||
AK088707.1 | BAC40517.1 | |||
AK134364.1 | BAE22116.1 | |||
AK150400.1 | BAE29526.1 | |||
AK150720.1 | BAE29798.1 | |||
AK150931.1 | BAE29968.1 | |||
AK151089.1 | BAE30102.1 | |||
AK151102.1 | BAE30112.1 | |||
AK151559.1 | BAE30503.1 | |||
AK151644.1 | BAE30574.1 | |||
AK152093.1 | BAE30941.1 | |||
AK152215.1 | BAE31043.1 | |||
AK152416.1 | BAE31201.1 | |||
AK152832.1 | BAE31532.1 | |||
AK159157.1 | BAE34862.1 | |||
AK165946.1 | BAE38478.1 | |||
AK167422.1 | BAE39510.1 | |||
AK168257.1 | BAE40205.1 | |||
AK177151.1 | ||||
AK181580.1 | ||||
AK212176.1 | ||||
BC015274.1 | AAH15274.1 | |||
BC024442.1 | AAH24442.1 | |||
Y11460.1 | CAA72246.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
