Assay Details
Target Gene Details
Entrez Gene ID: | 14049 |
Gene Name: | eyes absent 2 homolog (Drosophila) |
Gene Aliases: |
Eab1 |
Location: |
Chr.2:165595028-165771727 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eya2 | NM_001271962.1 | NP_001258891.1 | ||
NM_001271963.1 | NP_001258892.1 | |||
NM_010165.3 | NP_034295.1 | |||
AK137718.1 | BAE23473.1 | |||
AK141765.1 | BAE24826.1 | |||
AY212513.1 | ||||
BC003755.1 | AAH03755.1 | |||
U71208.1 | AAB51121.1 |
