Assay Details
Target Gene Details
Entrez Gene ID: | 98952 |
Gene Name: | family with sequence similarity 102, member A |
Gene Aliases: |
AI426465, C230093N12Rik, Eeig1 |
Location: |
Chr.2:32535359-32569750 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam102a | NM_153560.4 | 6 | 1272 | NP_705788.1 |
| AK049032.1 | 6 | 1276 | BAC33519.1 | |
| AK131145.1 | 5 | 2990 | BAD21395.1 | |
| AK157331.1 | 6 | 902 | BAE34050.1 | |
| AK157490.1 | 7 | 694 | BAE34101.1 | |
| AK158435.1 | 6 | 900 | ||
| AK165751.1 | 6 | 902 | ||
| AK170536.1 | 6 | 1300 | BAE41864.1 | |
| AK170847.1 | 6 | 893 | BAE42070.1 | |
| AK170951.1 | 6 | 899 | BAE42135.1 | |
| AK172536.1 | 6 | 899 | BAE43056.1 | |
| BC023752.1 | 6 | 717 | AAH23752.1 | |
| BC031157.1 | 6 | 535 | AAH31157.1 |

Genomic Map