Assay Details
Target Gene Details
Entrez Gene ID: | 30839 |
Gene Name: | F-box and WD-40 domain protein 5 |
Gene Aliases: |
AI159739, Fbw5 |
Location: |
Chr.2:25500767-25505471 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxw5 | NM_013908.4 | 3 | 359 | NP_038936.1 |
| AF176520.1 | 3 | 320 | AAF09130.1 | |
| AK019715.2 | 3 | 360 | BAB31841.1 | |
| AK154911.1 | 2 | 1298 | BAE32921.1 | |
| AK218179.1 | ||||
| BC010776.1 | 3 | 286 | AAH10776.1 |

Genomic Map