Assay Details
Target Gene Details
Entrez Gene ID: | 99326 |
Gene Name: | GTPase activating RANGAP domain-like 3 |
Gene Aliases: |
AW120551 |
Location: |
Chr.2:32986366-33131888 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Garnl3 | NM_178888.4 | 27 | 2687 | NP_849219.2 |
| AK034068.1 | 27 | 2689 | BAC28570.1 | |
| AK036258.1 | 26 | 2985 | ||
| AK083673.1 | 11 | 1259 | BAC38990.1 | |
| AK133163.1 | 28 | 2861 | BAE21537.1 | |
| BC132143.1 | 27 | 2630 |
More Information
Set Membership: |
|

Genomic Map