Assay Details
Target Gene Details
Entrez Gene ID: | 14415 |
Gene Name: | glutamate decarboxylase 1 |
Gene Aliases: |
EP10, GAD25, GAD44, GAD67, Gad-1 |
Location: |
Chr.2:70561763-70602014 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gad1 | NM_008077.4 | NP_032103.2 | ||
| AF326547.1 | AAL37393.1 | |||
| AF326548.1 | AAL37394.1 | |||
| AK054554.1 | BAC35823.1 | |||
| AK136801.1 | BAE23133.1 | |||
| AK148497.1 | BAE28589.1 | |||
| AK160868.1 | BAE36059.1 | |||
| BC027059.1 | AAH27059.1 | |||
| KM102984.1 | ||||
| Z49976.1 | CAA90277.1 |
Target Gene Details
Entrez Gene ID: | 68986 |
Gene Name: | glutamate decarboxylase 1, opposite strand |
Gene Aliases: |
1500002O10Rik |
Location: |
Chr.2:70489940-70563357 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gad1os | NR_040496.1 | |||
| AK005124.1 |
More Information
Set Membership: |
|

Genomic Map