Assay Details
Target Gene Details
Entrez Gene ID: | 381399 |
Gene Name: | BPI fold containing family B, member 4 |
Gene Aliases: |
Gm1006, Lplunc4 |
Location: |
Chr.2:153938217-153963852 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifb4 | NM_001034875.3 | 11 | 1509 | NP_001030047.2 |
| AK143420.1 | 8 | 873 | BAE25370.1 | |
| BC147610.1 | 8 | 872 | ||
| BC147625.1 | 8 | 872 |

Genomic Map