Assay Details
Target Gene Details
Entrez Gene ID: | 14571 |
Gene Name: | glycerol phosphate dehydrogenase 2, mitochondrial |
Gene Aliases: |
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38 |
Location: |
Chr.2:57237678-57370719 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpd2 | NM_001145820.1 | 11 | 1744 | NP_001139292.1 |
| NM_010274.3 | 11 | 1619 | NP_034404.3 | |
| AK034353.1 | 11 | 1528 | BAC28685.1 | |
| AK144716.1 | 11 | 1745 | BAE26028.1 | |
| AK151189.1 | 11 | 1532 | ||
| AK167152.1 | 11 | 1557 | BAE39294.1 | |
| BC021359.1 | 11 | 1713 | AAH21359.1 | |
| D50430.1 | 11 | 1521 | BAA08926.1 | |
| U60987.1 | 11 | 1555 | AAB50545.1 |
More Information
Set Membership: |
|

Genomic Map