Assay Details
Target Gene Details
Entrez Gene ID: | 14683 |
Gene Name: | GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus |
Gene Aliases: |
5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH |
Location: |
Chr.2:174283444-174346744 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
More Information
Set Membership: |
|

Genomic Map