Assay Details
Target Gene Details
Entrez Gene ID: | 76899 |
Gene Name: | golgi autoantigen, golgin subfamily a, 1 |
Gene Aliases: |
0710001G09Rik, 2210418B03Rik, AW107649, Golgi97, golgin-97 |
Location: |
Chr.2:39016155-39065579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golga1 | NM_001290649.1 | NP_001277578.1 | ||
| NM_029793.2 | NP_084069.1 | |||
| NR_110968.1 | ||||
| AK028516.1 | ||||
| AK044987.1 | ||||
| BC043090.1 | AAH43090.1 | |||
| BC053068.1 | AAH53068.1 | |||
| BC080733.1 | AAH80733.1 |

Genomic Map