Assay Details
Target Gene Details
Entrez Gene ID: | 76899 |
Gene Name: | golgi autoantigen, golgin subfamily a, 1 |
Gene Aliases: |
0710001G09Rik, 2210418B03Rik, AW107649, Golgi97, golgin-97 |
Location: |
Chr.2:39016155-39065579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Golga1 | NM_001290649.1 | NP_001277578.1 | ||
NM_029793.2 | NP_084069.1 | |||
NR_110968.1 | ||||
AK028516.1 | ||||
AK044987.1 | ||||
BC043090.1 | AAH43090.1 | |||
BC053068.1 | AAH53068.1 | |||
BC080733.1 | AAH80733.1 |
