Assay Details
Target Gene Details
Entrez Gene ID: | 227753 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF |
Location: |
Chr.2:35256359-35307902 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gsn | NM_001206367.1 | 3 | 328 | NP_001193296.1 |
NM_001206368.1 | 2 | 244 | NP_001193297.1 | |
NM_001206369.1 | 2 | 289 | NP_001193298.1 | |
NM_146120.4 | 2 | 351 | NP_666232.2 | |
AK076156.1 | 2 | 291 | BAC36223.1 | |
AK089934.1 | 3 | 283 | BAC41004.1 | |
AK135632.1 | 1 | 64 | ||
AK143664.1 | 2 | 318 | BAE25485.1 | |
AK146969.1 | 3 | 245 | BAE27574.1 | |
AK148957.1 | 3 | 308 | BAE28702.1 | |
AK150359.1 | 3 | 251 | BAE29494.1 | |
AK151684.1 | 3 | 270 | BAE30608.1 | |
AK153029.1 | 3 | 248 | BAE31661.1 | |
AK166831.1 | 2 | 264 | BAE39054.1 | |
AK166932.1 | 3 | 242 | BAE39126.1 | |
AK167918.1 | 3 | 258 | BAE39925.1 | |
AK168461.1 | 3 | 261 | BAE40358.1 | |
AK168532.1 | 3 | 262 | BAE40410.1 | |
AK168707.1 | 2 | 197 | BAE40549.1 | |
AK168725.1 | 3 | 245 | BAE40566.1 | |
AK168829.1 | 2 | 296 | ||
AK169041.1 | 3 | 248 | BAE40830.1 | |
AK169670.1 | 3 | 273 | BAE41291.1 | |
AK183123.1 | 3 | 293 | ||
AK186578.1 | 3 | 254 | ||
BC023143.1 | 2 | 289 | ||
BC060377.1 | 3 | 276 | AAH60377.1 | |
BY144206.1 | 2 | 289 | ||
J04953.1 | 1 | 138 | AAA37677.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
