Assay Details
Target Gene Details
Entrez Gene ID: | 71586 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
9130009C22Rik, Helicard, Hlcd, MDA5, RLR-2 |
Location: |
Chr.2:62595612-62646255 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifih1 | NM_001164477.1 | 2 | 831 | NP_001157949.1 |
| NM_027835.3 | 2 | 831 | NP_082111.2 | |
| AF374384.1 | 2 | 740 | AAM21359.1 | |
| AK037057.1 | 2 | 729 | BAC29687.2 | |
| AK040519.1 | 2 | 788 | BAC30614.1 | |
| AK149802.1 | 2 | 738 | BAE29095.1 | |
| AK153018.1 | 2 | 738 | BAE31652.1 | |
| AK156450.1 | 2 | 739 | BAE33716.1 | |
| AK168646.1 | 2 | 832 | BAE40503.1 | |
| AY075132.1 | 2 | 506 | AAL77205.1 | |
| BC080200.1 | 2 | 736 | AAH80200.1 | |
| EF113547.1 | 2 | 520 | ||
| EF113548.1 | 2 | 520 | ||
| EU084750.1 | 2 | 520 |

Genomic Map