Assay Details
Target Gene Details
Entrez Gene ID: | 16175 |
Gene Name: | interleukin 1 alpha |
Gene Aliases: |
Il-1a |
Location: |
Chr.2:129299609-129310901 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1a | NM_010554.4 | NP_034684.2 | ||
| AK156449.1 | BAE33715.1 | |||
| BC003727.1 | AAH03727.1 | |||
| X01450.1 | CAA25682.1 |
More Information
Set Membership: |
|

Genomic Map