Assay Details
Target Gene Details
Entrez Gene ID: | 23928 |
Gene Name: | laminin gamma 3 |
Gene Aliases: |
AI562206, AW240805 |
Location: |
Chr.2:31887281-31946535 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamc3 | NM_011836.3 | 12 | 2137 | NP_035966.2 |
| AF079520.1 | 12 | 2127 | AAD29851.1 | |
| AF083372.1 | 12 | 2074 | AAF08983.1 | |
| AK132948.1 | 9 | 1122 | ||
| BC096366.1 | 12 | 2137 | AAH96366.1 |
More Information
Set Membership: |
|

Genomic Map