Assay Details
Target Gene Details
Entrez Gene ID: | 16803 |
Gene Name: | lipopolysaccharide binding protein |
Gene Aliases: |
Bpifd2, Ly88 |
Location: |
Chr.2:158306493-158332852 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lbp | NM_008489.2 | 8 | 992 | NP_032515.2 |
| AA207419.1 | ||||
| AK143562.1 | 8 | 993 | ||
| BC004795.1 | 8 | 880 | AAH04795.1 | |
| X99347.1 | 8 | 942 | CAA67727.1 |

Genomic Map