Assay Details
Target Gene Details
Entrez Gene ID: | 71103 |
Gene Name: | glycosyltransferase 6 domain containing 1 |
Gene Aliases: |
4933411C14Rik, GT6m7 |
Location: |
Chr.2:25771820-25815940 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glt6d1 |
Target Gene Details
Entrez Gene ID: | 227630 |
Gene Name: | lipocalin 11 |
Gene Aliases: |
Gm109 |
Location: |
Chr.2:25777017-25780279 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lcn11 | NM_001100455.2 | NP_001093925.1 | ||
| AY301267.1 | AAQ81972.1 | |||
| BE864901.1 |
More Information
Set Membership: |
|

Genomic Map