Assay Details
Target Gene Details
Entrez Gene ID: | 19099 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 1 |
Gene Aliases: |
IB1, JIP-1, Jip1, Prkm8ip, Skip, mjip-2a |
Location: |
Chr.2:92383676-92401263 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapk8ip1 | NM_001202445.1 | NP_001189374.1 | ||
| NM_001202446.1 | NP_001189375.1 | |||
| NM_011162.5 | NP_035292.2 | |||
| AB221603.1 | ||||
| AF003115.1 | AAB66317.1 | |||
| AF054611.1 | AAD22580.1 | |||
| AF109768.1 | AAD38346.1 | |||
| AF109769.1 | AAD38347.1 | |||
| AF109770.1 | AAD38348.1 | |||
| AF109771.1 | AAD38349.1 | |||
| AF332075.1 | AAK56103.1 | |||
| AF332076.1 | AAK56104.1 | |||
| AK147033.1 | BAE27625.1 | |||
| AK192659.1 | ||||
| BC072578.1 | AAH72578.1 |
More Information
Set Membership: |
|

Genomic Map