Assay Details
Target Gene Details
Entrez Gene ID: | 668940 |
Gene Name: | myosin, heavy chain 7B, cardiac muscle, beta |
Gene Aliases: |
Myh14 |
Location: |
Chr.2:155611243-155634307 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 23 - Intron 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myh7b | NM_001085378.2 | NP_001078847.1 |
