Assay Details
Target Gene Details
Entrez Gene ID: | 17979 |
Gene Name: | nuclear receptor coactivator 3 |
Gene Aliases: |
2010305B15Rik, AW321064, Actr, Aib1, KAT13B, Rac3, Src3, Tram-1, Tram1, bHLHe42, p/Cip, pCip |
Location: |
Chr.2:165992637-166073242 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa3 | NM_008679.3 | 14 | 2793 | NP_032705.2 |
| AF000581.1 | 13 | 2685 | AAC05020.1 | |
| AK137517.1 | 3 | 461 | ||
| AK142306.1 | 14 | 2698 | BAE25022.1 | |
| AK156345.1 | 14 | 2770 | ||
| BC086482.1 | 14 | 2763 | ||
| BC093478.1 | 11 | 2424 | ||
| BC141177.1 | 13 | 2624 |
More Information
Set Membership: |
|

Genomic Map