Assay Details
Target Gene Details
Entrez Gene ID: | 29812 |
Gene Name: | N-myc downstream regulated gene 3 |
Gene Aliases: |
4833415O14Rik, AI747193, Ndr3 |
Location: |
Chr.2:156927342-156992111 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndrg3 | NM_013865.2 | NP_038893.1 | ||
| NM_180956.1 | NP_851287.1 | |||
| AB033922.1 | BAA85883.1 | |||
| AK002272.1 | ||||
| AK004689.1 | BAB23475.1 | |||
| AK029378.1 | ||||
| AK036277.1 | BAC29369.1 | |||
| AK037487.1 | BAC29818.1 | |||
| AK154927.1 | BAE32930.1 | |||
| BC018504.1 | AAH18504.1 | |||
| BY125191.1 |
More Information
Set Membership: |
|

Genomic Map