Assay Details
Target Gene Details
Entrez Gene ID: | 68349 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 3 |
Gene Aliases: |
0610010M09Rik, CI-30kD |
Location: |
Chr.2:90894636-90904721 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufs3 | NM_026688.2 | NP_080964.1 | ||
| AK002501.1 | BAB22149.1 | |||
| AK088337.1 | BAC40291.1 | |||
| AK160888.1 | 2 | 1633 | ||
| AK185633.1 | ||||
| AK206263.1 | ||||
| BC027270.1 | AAH27270.1 | |||
| BC119267.1 | ||||
| BC119269.1 |
More Information
Set Membership: |
|

Genomic Map