Assay Details
Target Gene Details
Entrez Gene ID: | 67134 |
Gene Name: | NOP56 ribonucleoprotein |
Gene Aliases: |
2310044F10Rik, Nol5a |
Location: |
Chr.2:130274367-130279313 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nop56 | NM_024193.2 | 5 | 529 | NP_077155.2 |
| AK009799.1 | 5 | 515 | BAB26511.1 | |
| AK011481.1 | 5 | 496 | BAB27647.3 | |
| AK011744.1 | 5 | 494 | ||
| AK013528.1 | 5 | 496 | ||
| AK030730.1 | 4 | 649 | ||
| AK077795.1 | 5 | 496 | BAC37015.1 | |
| AK139924.1 | 5 | 496 | BAE24185.1 | |
| AK146243.1 | 5 | 496 | BAE27007.1 | |
| AK150258.1 | 5 | 496 | BAE29417.1 | |
| AK154024.1 | 5 | 497 | ||
| BC021355.1 | 5 | 491 | AAH21355.1 |
More Information
Set Membership: |
|

Genomic Map