Assay Details
Target Gene Details
Entrez Gene ID: | 67134 |
Gene Name: | NOP56 ribonucleoprotein |
Gene Aliases: |
2310044F10Rik, Nol5a |
Location: |
Chr.2:130274367-130279313 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nop56 | NM_024193.2 | NP_077155.2 | ||
| AK009799.1 | BAB26511.1 | |||
| AK011744.1 | ||||
| AK013528.1 | ||||
| AK030730.1 | 5 | 1937 | ||
| AK037405.1 | ||||
| AK077795.1 | BAC37015.1 | |||
| AK139924.1 | BAE24185.1 | |||
| AK146243.1 | BAE27007.1 | |||
| AK150258.1 | BAE29417.1 | |||
| AK154024.1 | 6 | 1785 | ||
| BC002231.1 | AAH02231.1 | |||
| BC021355.1 | AAH21355.1 |
More Information
Set Membership: |
|

Genomic Map