Assay Details
Target Gene Details
Entrez Gene ID: | 59126 |
Gene Name: | NIMA (never in mitosis gene a)-related expressed kinase 6 |
Gene Aliases: |
1300007C09Rik |
Location: |
Chr.2:38511697-38587490 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nek6 | NM_001159631.1 | 9 | 907 | NP_001153103.1 |
| NM_021606.3 | 9 | 900 | NP_067619.1 | |
| AF218847.1 | 9 | 902 | AAG16653.1 | |
| AK004925.1 | 9 | 877 | BAB23676.2 | |
| AK011519.1 | 9 | 886 | BAB27673.2 | |
| AK018168.1 | 9 | 889 | ||
| AK029266.1 | 9 | 905 | ||
| AK075717.1 | 9 | 874 | BAC35907.1 | |
| AK075836.1 | 9 | 886 | BAC35995.1 | |
| AK089919.1 | 9 | 861 | BAC40995.1 | |
| AK170757.1 | 9 | 907 | BAE42008.1 | |
| AK185671.1 | 2 | 168 | ||
| BC019524.1 | 9 | 969 | AAH19524.1 |
More Information
Set Membership: |
|

Genomic Map