Assay Details
Target Gene Details
Entrez Gene ID: | 228866 |
Gene Name: | PDX1 C-terminal inhibiting factor 1 |
Gene Aliases: |
2310022K11Rik, C20orf67, F730014I05Rik |
Location: |
Chr.2:164879368-164891437 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcif1 | NM_146129.3 | NP_666241.1 | ||
| AK089358.1 | BAC40853.1 | |||
| AK150740.1 | BAE29814.1 | |||
| BC031431.1 | AAH31431.1 |

Genomic Map