Assay Details
Target Gene Details
Entrez Gene ID: | 26428 |
Gene Name: | origin recognition complex, subunit 4 |
Gene Aliases: |
Orc4P, Orc4l, mMmORC4 |
Location: |
Chr.2:48902754-48950272 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orc4 | NM_011958.3 | 11 | 1068 | NP_036088.3 |
| NR_033442.1 | ||||
| AJ003140.1 | 11 | 1029 | CAA05896.1 | |
| AK010258.1 | 11 | 1052 | BAB26801.1 | |
| AK011550.1 | BAC25339.1 | |||
| AK076324.1 | 11 | 1069 | BAC36300.1 | |
| AK084326.1 | 11 | 1037 | BAC39161.1 | |
| AK145454.1 | 11 | 1080 | ||
| AK169291.1 | 11 | 1052 | BAE41048.1 | |
| BC015072.1 | 11 | 1040 | AAH15072.1 | |
| BC019748.1 | 11 | 1010 | AAH19748.1 | |
| Y16386.1 | 10 | 900 | CAA76188.1 |
More Information
Set Membership: |
|

Genomic Map