Assay Details
Target Gene Details
Entrez Gene ID: | 26428 |
Gene Name: | origin recognition complex, subunit 4 |
Gene Aliases: |
Orc4P, Orc4l, mMmORC4 |
Location: |
Chr.2:48902754-48950272 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orc4 | NM_001177313.1 | NP_001170784.1 | ||
| NM_011958.3 | NP_036088.3 | |||
| NR_033442.1 | ||||
| AJ003140.1 | CAA05896.1 | |||
| AK010258.1 | BAB26801.1 | |||
| AK011550.1 | BAC25339.1 | |||
| AK012669.1 | BAB28396.1 | |||
| AK049534.1 | ||||
| AK076324.1 | BAC36300.1 | |||
| AK084326.1 | BAC39161.1 | |||
| AK145454.1 | ||||
| AK169291.1 | BAE41048.1 | |||
| BC015072.1 | AAH15072.1 | |||
| BC019748.1 | AAH19748.1 | |||
| Y16386.1 | CAA76188.1 |
More Information
Set Membership: |
|

Genomic Map